Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OGFOD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562549 Supplier Invitrogen™ Supplier No. PA562549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGVTDVVITR EEAERIRSVA EKGLSLGGSD GGASILDLHS GALSVGKHFV NLYRYFGDKI QNIFSEEDFR LYREVRQKVQ LTIAEAFGIS ASSLHLTKPT FFSRINSTEA RTAHDEYWHA HVDKVTYGS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

OGFOD3 gene ontology annotations related to this gene include dioxygenase activity; iron ion binding; L-ascorbic acid binding; oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OGFOD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OGFOD3
Gene Accession No. Q6PK18
Gene Alias 1110031I02Rik; 2-oxoglutarate and iron dependent oxygenase domain containing 3; 2-oxoglutarate and iron-dependent oxygenase domain containing 3; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 3; C17orf101; Ogfod3; PKHD domain-containing transmembrane protein C17orf101; PKHD domain-containing transmembrane protein C17orf101 homolog; RGD1305007
Gene Symbols OGFOD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OGFOD3. Recombinant protein control fragment (Product #RP-89703).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.