Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGG1 Antibody (2B4), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252724APC
Description
OGG1 Monoclonal specifically detects OGG1 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
OGG1 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
OGG1 | |
Partial recombinant protein made to an internal sequence of the human OGG1 protein (between amino acids 100-300) [Uniprot: O15527] | |
0.1 mL | |
Base Excision Repair, Cancer, Chromatin Research, DNA Repair | |
4968 | |
Store at 4C in the dark. |
Flow Cytometry | |
2B4 | |
Flow Cytometry, Flow (Intracellular) | |
8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction