Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGG1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100106MFV450
Description
OGG1 Polyclonal antibody specifically detects OGG1 in Human, Mouse, Rat, Porcine, Primate, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
OGG1 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt# O15527] | |
0.1 mL | |
Base Excision Repair, Cancer, Chromatin Research, DNA Repair | |
4968 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Primate, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction