Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGG1 Mouse anti-Human, Clone: CPTC-OGG1-1, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | OGG1 |
---|---|
Clone | CPTC-OGG1-1 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
OGG1 Monoclonal specifically detects OGG1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
OGG1 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Base Excision Repair, Cancer, Chromatin Research, DNA Repair | |
10 mM PBS with 0.05% BSA | |
4968 | |
IgG2c κ | |
Protein A or G purified |
CPTC-OGG1-1 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
Recombinant full-length human OGG1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title