Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGT/O-GlcNAc transferase Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $651.90
Specifications
| Antigen | OGT/O-GlcNAc transferase |
|---|---|
| Concentration | Lot-Specific |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3341
![]()
|
Active Motif
61356 |
10 μL |
Each for $159.90
|
|
|||||
50-199-3340
![]()
|
Active Motif
61355 |
100 μL |
Each for $651.90
|
|
|||||
Description
O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.Specifications
| OGT/O-GlcNAc transferase | |
| ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| GlcNAc | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Lot-Specific | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| This antibody was raised against a recombinant protein corresponding to the N-terminal region of human OGT. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title