Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Measurement Skills Assessment Review Kit
Shop All OHAUS Products

Click to view available options
:
Ohaus Measurement Mastery Game
Description
Includes
- Set of 750 standards-based assessment review questions
- 12 game boards with materials for 36 students to play games simultaneously
- CD-ROM with over 700 interactive assessment questions
- Comprehensive teacher planning and resource guide
- Free 90-day online subscription to NewPath Learning™ that includes thousands of resources
Provides coverage of up-to-date measurement concepts with easy-to-follow instructions and minimal setup.
- Specifically developed to provide comprehensive coverage of standards-based measurement topics
- Differentiated instruction capabilities accommodate visual, tactile, oral and aural learning styles
- Flexible design accommodates lesson times from 5 to 30 minutes
- Standard rules and game board configuration maximize productive learning time
- All questions are grade-appropriate and leveled by content and readability
- Enhance the experience with hands-on activities incorporating the Harvard Junior™ mechanical balance (sold separately)
- Class size: 36 students

Specifications
Specifications
Product Type | Measurement Assessment Puzzle |
Grade | 3 to 5 |
Class Size | 36 Students |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction