Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Starter Series Electrode: ST260 2-in-1 pH Electrode
SCI_ED

Catalog No. s14235
Click to view available options
Connector Types:
BNC

Use in combination with Starter bench and portable meters. OHAUS Starter Series Electrode: ST260 2-in-1 pH Electrodes need to work with STTEMP30 temperature electrodes for 0.01 pH accuracy. ST260 is fit for samples which may react with Ag+ ions such as tri-buffer solution.

  • Designed to produce exact results time and time again
  • Sturdy and durable
  • Constructed from glass shafts and built to withstand daily use
  • Includes a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements
  • Fits in electrode holders on bench meters and electrode clips on all portable meters
  • Reference System: double salt bridge, Ag/AgCl-KCl
  • Salt Bridge Solution: saturated Pure KCl solution
  • Reference Electrolyte: 3M KCl Solution

Specifications

  • Parameters Measured: pH
  • pH Range: 0.00 to 14.00
  • Temperature Range: 0° to 100°C
  • Connection: BNC
  • Cable Length: 1m
  • Shaft dia. x L: 12 x 120mm
  • Zero Potential: 7.00±0.25 pH
  • Slope: ≥97%
  • Impedance: ≤250MOhm
  • Reference Diaphragm: Sleeve

Applications

Ideal for use in lab or in the field.

TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Junction 3M KCl Solution
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0 to 100°
Type 2-in-1 pH Electrode
Filling Solution Type Salt Bridge Solution: saturated Pure KCl solution; Reference Electrolyte: 3M KCl Solution
For Use With (Application) Samples which may react with Ag+ ions such as tri-buffer solution.
For Use With (Equipment) Starter bench and Portable meters
Internal Reference Double salt bridge, Ag/AgCl-KCl
Model ST260
Temperature (Metric) Range 0°C to 100°C
Length (Metric) Cable 1 m
Length (Metric) Stem 120 mm
Diameter (Metric) Body 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.