Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OIF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989047
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9047 Supplier Bioss Supplier No. BS9407R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.

Specifications

Antigen OIF
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene OGN
Gene Accession No. P20774
Gene Alias 3110079A16Rik; BOS_9223; corneal keratan sulfate proteoglycan; Corneal keratan sulfate proteoglycan 25 core protein; KSPG25 protein; Mimecan; mimecan proteoglycan; mimican; Og; Ogn; OIF; Osteoglycin; osteoglycin (osteoinductive factor); osteoglycin (osteoinductive factor, mimecan); osteoglycin preproprotein; Osteoinductive factor; SLRR3A
Gene Symbols OGN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OIF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4969
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.