Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OIP106 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | OIP106 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
OIP106 Polyclonal specifically detects OIP106 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
OIP106 | |
Polyclonal | |
Rabbit | |
Human | |
106 kDa O-GlcNAc transferase-interacting protein, KIAA1042OGT(O-Glc-NAc transferase)-interacting protein 106 KDa, OIP106OGT(O Glc NAc transferase) interacting protein 106 KDa, trafficking kinesin-binding protein 1, trafficking protein, kinesin binding 1 | |
TRAK1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
22906 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CTPRRLSLAESFTNTRESTTTMSTSLGLVWLLKERGISAAVYDPQSWDRAGRGSLLHSYTPKMAVIPSTPPNSPMQTPTSSPPSFEFKCTSPPYDNFLASKPASSILREVREKNVRSSESQTDVSVSNLNLVD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title