Learn More
OIP5 Monoclonal Antibody (OTI1F1)

Description
OIP5 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | OIP5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1F1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | OIP5 |
| Gene Accession No. | O43482 |
| Gene Alias | 5730547N13Rik; cancer/testis antigen 86; CT86; hMIS18beta; LAP2alpha interactor-25; LINT-25; MIS18 kinetochore protein homolog B; MIS18B; MIS18beta; OIP5; OIP-5; Opa interacting protein 5; opa-interacting protein 5; Protein Mis18-beta; RGD1564263 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.