Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OIP5 Mouse anti-Human, Clone: 1E9B5, Proteintech
SDP

Catalog No. 501736887 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6887 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6887 Supplier Proteintech Group Inc Supplier No. 664611IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.

Specifications

Antigen OIP5
Applications Western Blot
Classification Monoclonal
Clone 1E9B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene OIP5
Gene Accession No. O43482
Gene Alias 5730547N13Rik, Cancer/testis antigen 86, CT86, LINT 25, MIS18B, MIS18beta, OIP 5, OIP5, Opa interacting protein 5, Protein Mis18 beta
Gene Symbols OIP5
Host Species Mouse
Immunogen OIP5 Fusion Protein Ag2790
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11339
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.