Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OIP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562579 Supplier Invitrogen™ Supplier No. PA562579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CGSCGIPVGF HLYSTHAALA ALRGHFCLSS DKMVCYLLKT KAIVNASEMD IQNVPLSEKI AELKEKIVLT HNRLKSLMKI LSEVT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OIP5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OIP5
Gene Accession No. O43482
Gene Alias 5730547N13Rik; cancer/testis antigen 86; CT86; hMIS18beta; LAP2alpha interactor-25; LINT-25; MIS18 kinetochore protein homolog B; MIS18B; MIS18beta; OIP5; OIP-5; Opa interacting protein 5; opa-interacting protein 5; Protein Mis18-beta; RGD1564263
Gene Symbols OIP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OIP5. Recombinant protein control fragment (Product #RP-98056).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11339
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.