Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OIP5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | OIP5 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7745
![]() ![]() |
Proteintech Group Inc
121421AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.Specifications
OIP5 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O43482 | |
11339 | |
OIP5 Fusion Protein Ag2790 | |
Primary | |
-20°C | |
OIP5 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
5730547N13Rik, Cancer/testis antigen 86, CT86, LINT 25, MIS18B, MIS18beta, OIP 5, OIP5, Opa interacting protein 5, Protein Mis18 beta | |
OIP5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title