Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OIT3 Polyclonal Antibody

Catalog No. PIPA5121728
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121728 20 μL
PIPA5119319 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121728 Supplier Invitrogen™ Supplier No. PA5121728
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OIT3 Polyclonal Antibody for Western Blot, IHC (P)

May be involved in hepatocellular function and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OIT3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene OIT3
Gene Accession No. Q8R4V5, Q8WWZ8
Gene Alias EF-9; liver-specific zona pellucida domain-containing protein; liver-specific ZP domain-containing protein; LZP; OIT3; oncoprotein induced transcript 3; oncoprotein-induced transcript 3 protein; UNQ826/PRO1753
Gene Symbols OIT3
Host Species Rabbit
Immunogen Recombinant protein Oncoprotein Induced Transcript 3. The antigen corresponds to amino acid range 297-506 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170392, 18302
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.