Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OIT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | OIT3 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119320
|
Invitrogen™
PA5119320 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121729
|
Invitrogen™
PA5121729 |
20 μL |
Each for $237.00
|
|
|||||
Description
OIT3 Polyclonal Antibody for Western Blot, IHC (P)
May be involved in hepatocellular function and development.Specifications
OIT3 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
OIT3 | |
EF-9; liver-specific zona pellucida domain-containing protein; liver-specific ZP domain-containing protein; LZP; OIT3; oncoprotein induced transcript 3; oncoprotein-induced transcript 3 protein; UNQ826/PRO1753 | |
OIT3 | |
Primary | |
Antigen affinity chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q6V0K7 | |
294559 | |
Recombinant protein Oncoprotein Induced Transcript 3. The antigen corresponds to amino acid range 251-524 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title