Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLA1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501570478
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0478 Supplier Bethyl Laboratories, Inc Supplier No. A305459A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan

GTPBP9 belongs to the GTP1/OBG family and the function remains unknown.

Specifications

Antigen OLA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene OLA1
Gene Accession No. Q9CZ30, Q9NTK5
Gene Alias DOC45, GBP45, GTBP9, GTPBP9, DNA damage-regulated overexpressed in cancer 45 protein, GTP-binding protein 9 (putative), GTP-binding protein PTD004, homologous yeast-44.2 protein, obg-like ATPase 1
Gene Symbols Ola1
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29789, 67059
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.