Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Olaquindox Monoclonal Antibody (3F2)
GREENER_CHOICE

Catalog No. PIMA549668
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549668 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549668 Supplier Invitrogen™ Supplier No. MA549668
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Olaquindox is a chemical compound that has been used as a feed additive in livestock production to promote growth and prevent diseases. Olaquindox is a gene target that encodes for a protein involved in the metabolism and detoxification of olaquindox in animals. This gene is important for understanding the mechanisms of olaquindox toxicity and resistance in livestock and for developing strategies to mitigate the potential health and environmental risks associated with the use of this compound in animal feed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olaquindox
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3F2
Concentration 6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of OLA hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.