Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ OLDA
GREENER_CHOICESDP

Catalog No. 16415 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-415 5 mg
16-412-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-415 Supplier R&D Systems™ Supplier No. 1641/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective endogenous TRPV1 agonist

Potent endogenous vanilloid TRPV1 (VR1) receptor agonist (EC50 = 36 nM at hVR1) with low affinity for rCB1 receptors (Ki = 1.6 μM). Potently induces VR1-mediated thermal hyperalgesia in rats in vivo.

Chemical Identifiers

CAS 105955-11-1
Molecular Formula C26H43NO3

Specifications

Quantity 5 mg
Product Line Tocris
Formula Weight 417.63
Purity >99%
Molecular Weight (g/mol) 417.634
Chemical Name or Material OLDA
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.