Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oleamide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Endogenous sleep-inducing lipid. Acts as an agonist at the CB1 cannabinoid receptor (EC50 = 1.64 μM). Also appears to potentiate the actions of 5-HT on 5-HT2A and 2C receptors, and act via an allosteric regulatory site on 5-HT7 receptors.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 281.48 |
Percent Purity | >98% |
Chemical Name or Material | Oleamide |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction