Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleamide, Tocris Bioscience™

Catalog No. 087810
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Sleep-inducing brain lipid

Endogenous sleep-inducing lipid. Acts as an agonist at the CB1 cannabinoid receptor (EC50 = 1.64 μM). Also appears to potentiate the actions of 5-HT on 5-HT2A and 2C receptors, and act via an allosteric regulatory site on 5-HT7 receptors.

Chemical Identifiers

CAS 301-02-0
Molecular Formula C18H35NO
Molecular Weight (g/mol) 281.484
InChI Key FATBGEAMYMYZAF-KTKRTIGZSA-N
Synonym oleamide, oleic acid amide, oleylamide, oleyl amide, adogen 73, crodamide o, slip-eze, armoslip cp, crodamide or, cis-9,10-octadecenoamide
PubChem CID 5283387
ChEBI CHEBI:116314
IUPAC Name (Z)-octadec-9-enamide
SMILES CCCCCCCCC=CCCCCCCCC(=O)N

Specifications

Quantity 10 mg
Formula Weight 281.48
Percent Purity >98%
Chemical Name or Material Oleamide
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.