Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic acid, 97%

Catalog No. p-3737122
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 112-80-1 | C18H34O2 | 282.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (Z)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Melting Point 13.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 360.0°C
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH=CH(CH2)7COOH
Refractive Index 1.4585 to 1.4605
Quantity 5 g
Beilstein 02,463
Fieser 01,759
Merck Index 15,6921
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether,, fixed and volatile oils
Viscosity 39.1 mPa.s (20°C)
Formula Weight 282.46
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Oleic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 204-007-1
Recommended Storage −20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.