Learn More
Oleic acid, tech. 90%
Description
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.
Solubility
Miscible with ethanol, ether, acetone, chloroform, dimethyl formamide and dimethyl sulfoxide.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and aluminum.

Specifications
Specifications
Melting Point | 13°C to 14°C |
Density | 0.887 |
Boiling Point | 286°C (100 mmHg) |
Assay Percent Range | 90% |
Refractive Index | 1.461 |
Quantity | 5000 mL |
Beilstein | 1726542 |
Sensitivity | Air sensitive |
Merck Index | 14,6828 |
Solubility Information | Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide. |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement P280-P305+P351+P338-P362-P321-P332+P313-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse. Specific treatment (see label). If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. |
EINECSNumber | 204-007-1 |
RTECSNumber | RG2275000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.