Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic acid, tech. 90%

Catalog No. aaa16663au
Click to view available options
Quantity:
1000 mL
5000 mL

CAS: 112-80-1 | C18H34O2 | 282.47 g/mol

Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.

Solubility
Miscible with ethanol, ether, acetone, chloroform, dimethyl formamide and dimethyl sulfoxide.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Melting Point 13°C to 14°C
Density 0.887 g/mL
Boiling Point 286°C (100 mmHg)
Assay Percent Range 90%
Refractive Index 1.461
Quantity 1000 mL
Beilstein 1726542
Sensitivity Air sensitive
Merck Index 14,6828
Solubility Information Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide.
IUPAC Name (9E)-octadec-9-enoic acid
Formula Weight 282.47
Percent Purity 90%
Grade Technical
Chemical Name or Material Oleic Acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P362-P321-P332+P313-P337+P313-P302+P352
EINECSNumber 204-007-1
RTECSNumber RG2275000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.