Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic Anhydride 95.0+%, TCI America™
SDP

Catalog No. O02271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02271G 1 g
O02275G 5 g
O022725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O02271G Supplier TCI America Supplier No. O02271G

Chemical Identifiers

CAS 24909-72-6
Molecular Formula C36H66O3
Molecular Weight (g/mol) 546.92
MDL Number MFCD00010459
InChI Key OCNZHGHKKQOQCZ-CLFAGFIQSA-N
Synonym cis-9-Octadecenoic Anhydride
PubChem CID 5369123
IUPAC Name (9Z)-octadec-9-enoyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 546.92
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Oleic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.