Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl alcohol, 99+%

Catalog No. AAH3347506
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 143-28-2 | C18H36O | 268.49 g/mol

Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

Solubility
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Melting Point ∼6°C
Density 0.849
Boiling Point 206°C to 207°C (13 mmHg)
Flash Point 180°C (356°F)
Refractive Index 1.46
Quantity 5 g
Beilstein 1723962
Solubility Information Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 268.48
Percent Purity ≥99%
Chemical Name or Material Oleyl alcohol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315
Causes skin irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 205-597-3
RTECSNumber RG4120000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.