Learn More
Invitrogen™ OLFM1 Polyclonal Antibody

Description
Immunogen sequence: IEVLDRRTQR DLQYVEKMEN QMKGLESKFK QVEESHKQHL ARQFKAIKAK MDELRPLIPV LEEYKADAKL VLQFKEEVQN LTSVLNELQE EIGAYDYDEL QSRVSNLEER LRACMQKLAC GKLTGISDPV T; Positive Samples: Mouse brain, MCF7, A-549, SW480; Cellular Location: Cell junction, Cell projection, Endoplasmic reticulum, Perikaryon, axon, synapse.
Specifications
Specifications
| Antigen | OLFM1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | OLFM1 |
| Gene Accession No. | O88998, Q99784 |
| Gene Alias | 1B426B; AMY; AW742568; D2Sut1e; neuroblastoma protein; neuronal olfactomedin-related ER localized protein; Noe1; Noel; Noel1; Noelin; Noelin 1; Noelin 2; NOELIN1; OlfA; olfactomedin 1; olfactomedin related ER localized protein; olfactomedin-1; OLFM1; Pancortin; pancortin 1; Pancortin 1-4; Pancortin3 |
| Gene Symbols | OLFM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.