Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548087
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548087 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548087 Supplier Invitrogen™ Supplier No. PA548087
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) Olfactomedin-1 is observed and less than 5% cross-reactivity with recombinant human Olfactomedin-4 and rmOlfactomedin-L3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Involved in transforming growth factor beta (TGF-beta)-induced smooth muscle differentiation. TGF-beta induces expression and nuclear translocation of OLFM2 where it binds to SRF, causing its dissociation from the transcriptional repressor HEY2/HERP1 and facilitating binding of SRF to target genes. Plays a role in AMPAR complex organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFM2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene OLFM2
Gene Accession No. O95897
Gene Alias A030009A06Rik; neuronal olfactomedin related ER localized protein 2; NOE2; noelin 2; NOELIN2; Noelin-2; NOELIN2_V1; olfactomedin 2; olfactomedin-2; OlfC; OLFM2
Gene Symbols OLFM2
Host Species Sheep
Immunogen CHO-derived recombinant human Olfactomedin-2/Noelin-2 Gln21-Pro454.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93145
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.