Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565590 Supplier Invitrogen™ Supplier No. PA565590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLFYNKYQSNV VVKYHFRSRS VLVQRSLPGA GYNNTFPYSW GGFSDMDFMV DES Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Involved in transforming growth factor beta (TGF-beta)-induced smooth muscle differentiation. TGF-beta induces expression and nuclear translocation of OLFM2 where it binds to SRF, causing its dissociation from the transcriptional repressor HEY2/HERP1 and facilitating binding of SRF to target genes. Plays a role in AMPAR complex organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFM2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OLFM2
Gene Accession No. O95897
Gene Alias A030009A06Rik; neuronal olfactomedin related ER localized protein 2; NOE2; noelin 2; NOELIN2; Noelin-2; NOELIN2_V1; olfactomedin 2; olfactomedin-2; OlfC; OLFM2
Gene Symbols OLFM2
Host Species Rabbit
Immunogen Recombinant Human OLFM2. Recombinant protein control fragment (Product #RP-98972).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.