Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLFM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | OLFM2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7627
![]() ![]() |
Proteintech Group Inc
119971AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Involved in transforming growth factor beta (TGF-beta)-induced smooth muscle differentiation. TGF-beta induces expression and nuclear translocation of OLFM2 where it binds to SRF, causing its dissociation from the transcriptional repressor HEY2/HERP1 and facilitating binding of SRF to target genes. Plays a role in AMPAR complex organization.Specifications
OLFM2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O95897, Q568Y7, Q8BM13 | |
244723, 313783, 93145 | |
OLFM2 Fusion Protein Ag2642 | |
Primary | |
-20°C | |
OLFM2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
NOE2, Noelin 2, NOELIN2, NOELIN2_V1, olfactomedin 2, OlfC, OLFM2 | |
Olfm2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title