Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFM3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550866

Catalog No. PA550866


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total OLFM3 protein.

OLFM3 (olfactomedin 3), also known as NOE3, is a 478 amino acid protein that interacts with myocilin. Myocilin is an extracellular protein that plays a key role in the actomyosin system and is responsible for controlling intraocular pressure. OLFM3 is a secreted protein that contains an olfactomedin-like (OLF) domain, an approximately 260 amino acid motif commonly found in secreted glycoproteins. OLFM3 localizes to the Golgi apparatus of the cell and is highly expressed in both eye and brain tissue. Mutations in the gene that encodes OLFM3 may cause severe glaucoma, a condition in which increased intraocular pressure within the eyeball causes loss of eye sight.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OLFM3
Polyclonal
Unconjugated
Olfm3
B230206G02Rik; NOE3; NOELIN3; Noelin-3; olfactomedin 3; olfactomedin related ER localized protein 3; olfactomedin-3; OLFM3; OPTIMEDIN; UNQ1924/PRO4399
Rabbit
Antigen affinity chromatography
RUO
118427, 229759
-20°C
Liquid
Western Blot
1.1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P63056, Q96PB7
Olfm3
Fusion protein corresponding to residues near the C terminal of human olfactomedin 3.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.