Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110059 Supplier Invitrogen™ Supplier No. PA5110059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HQRVLSLETR LRDCMKKLTC GKLMKITGPI TVKTSGTRFG AWMTDPLASE KNNRVWYMDS YTNNKIVREY KSIADFVSGA ESRTYNLPFK WAGTNHVVYN G.

OLFM3 (olfactomedin 3), also known as NOE3, is a 478 amino acid protein that interacts with myocilin. Myocilin is an extracellular protein that plays a key role in the actomyosin system and is responsible for controlling intraocular pressure. OLFM3 is a secreted protein that contains an olfactomedin-like (OLF) domain, an approximately 260 amino acid motif commonly found in secreted glycoproteins. OLFM3 localizes to the Golgi apparatus of the cell and is highly expressed in both eye and brain tissue. Mutations in the gene that encodes OLFM3 may cause severe glaucoma, a condition in which increased intraocular pressure within the eyeball causes loss of eye sight.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFM3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Olfm3
Gene Accession No. P63056, P63057, Q96PB7
Gene Alias B230206G02Rik; NOE3; NOELIN3; Noelin-3; olfactomedin 3; olfactomedin related ER localized protein 3; olfactomedin-3; OLFM3; OPTIMEDIN; UNQ1924/PRO4399
Gene Symbols Olfm3
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118427, 229759, 252920
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.