Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLFM4 Antibody (12), PE , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306415PE
Description
OLFM4 Monoclonal antibody specifically detects OLFM4 in Human samples. It is validated for Flow CytometrySpecifications
OLFM4 | |
Monoclonal | |
PE | |
Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLM4, UNQ362 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OLFM4 (NP_006409.3; Met 1-Gln 510). | |
0.1 mL | |
Cancer | |
10562 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
12 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction