Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLFM4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224535SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224535SS 0.025 mg
NBP224535 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224535SS Supplier Novus Biologicals Supplier No. NBP224535SS

Rabbit Polyclonal Antibody has been used in 4 publications

OLFM4 Polyclonal specifically detects OLFM4 in Human, Mouse, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Single Cell Western.

Specifications

Antigen OLFM4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-3 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 12 ug/ml, Immunohistochemistry-Paraffin 1-5 ug/ml, Single Cell Western 100 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q6UX06
Gene Alias Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLM4, UNQ362
Gene Symbols OLFM4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids between 400 and 450 of human OLFM4 was used as immunogen for this antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10562
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoform CRA_a) and chimpanzee, 85% in monkey, mouse and rat, 76% in cow and 75% homologous in dog.
Target Species Human, Mouse, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.