Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLFM4 Antibody (SR1959), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321911100UL
Description
OLFM4 Monoclonal antibody specifically detects OLFM4 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
OLFM4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1959 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLM4, UNQ362 | |
A synthesized peptide derived from human OLFM4 (Uniprot #: Q6UX06) | |
100 μg | |
Cancer | |
10562 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction