Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OLFML1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113011
Description
OLFML1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The specific function of this protein remains unknown.
Specifications
OLFML1 | |
Polyclonal | |
Unconjugated | |
OLFML1 | |
6720478C22; AV321997; BC047207; mONT2; MVAL564; olfactomedin like 1; olfactomedin-like 1; olfactomedin-like protein 1; OLFML1; ONT2; UNQ564; UNQ564/PRO1126 | |
Rabbit | |
Protein G | |
RUO | |
244198, 283298 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q6UWY5 | |
OLFML1 | |
Recombinant Human Olfactomedin-like protein 1 protein (201-402AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction