Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLFML2A Polyclonal Antibody
OLFML2A Polyclonal Antibody
Supplier: Thermo Scientific PA554343
Description
OLFML2A Polyclonal Antibody for ICC/IF, IHC

Specifications
OLFML2A | |
Polyclonal | |
Unconjugated | |
OLFML2A | |
4932431K08Rik; mFLJ00237; olfactomedin like 2A; olfactomedin-like 2A; olfactomedin-like protein 2A; OLFML2A; Photomedin-1; PRO34319; UNQ9394/PRO34319 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
169611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68BL7 | |
OLFML2A | |
Recombinant protein corresponding to Human OLFML2A. Recombinant protein control fragment (Product #RP-93075). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction