Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFML2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554577 Supplier Invitrogen™ Supplier No. PA554577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FYTVETVSSG TDCRCSCTAP PSSLNPCENE WKMEKLKKQA PELLKLQSMV DLLEGTLYSM DLMKVHAYVH KVASQMNTLE ESIKANLSRE N Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.

OLFML2A contains one olfactomedin-like domain. Its exact function is unknown. There are three named isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFML2A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OLFML2A
Gene Accession No. Q68BL7
Gene Alias 4932431K08Rik; mFLJ00237; olfactomedin like 2A; olfactomedin-like 2A; olfactomedin-like protein 2A; OLFML2A; Photomedin-1; PRO34319; UNQ9394/PRO34319
Gene Symbols OLFML2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OLFML2A. Recombinant protein control fragment (Product #RP-93076).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.