Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFML2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599687 Supplier Invitrogen™ Supplier No. PA599687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Photomedin-2.

This gene encodes an olfactomedin domain-containing protein. Most olfactomedin domain-containing proteins are secreted glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFML2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OLFML2B
Gene Accession No. Q3V1G4, Q68BL8
Gene Alias 1110018N05Rik; 4832415H08Rik; AI467542; olfactomedin like 2B; olfactomedin-like 2B; olfactomedin-like protein 2B; Olfml2b; Photomedin-2
Gene Symbols OLFML2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human OLFML2B(Accession Q68BL8), corresponding to amino acid residues K172-M222.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25903, 320078
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.