Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFML2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113006
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113006 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113006 Supplier Invitrogen™ Supplier No. PA5113006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OLFML2B Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes an olfactomedin domain-containing protein. Most olfactomedin domain-containing proteins are secreted glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFML2B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OLFML2B
Gene Accession No. Q3V1G4, Q68BL8
Gene Alias 1110018N05Rik; 4832415H08Rik; AI467542; olfactomedin like 2B; olfactomedin-like 2B; olfactomedin-like protein 2B; Olfml2b; Photomedin-2
Gene Symbols OLFML2B
Host Species Rabbit
Immunogen Recombinant Human Olfactomedin-like protein 2B protein (451-750AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25903, 320078
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.