Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLFML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113463 Supplier Invitrogen™ Supplier No. PA5113463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OLFML3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the olfactomedin-like gene family which also includes genes encoding noelin, tiarin, myocilin, amassin, optimedin, photomedin, and latrophilin. The encoded protein is a secreted extracellular matrix glycoprotein with a C-terminal olfactomedin domain that facilitates protein-protein interactions, cell adhesion, and intercellular interactions. It serves as both a scaffold protein that recruits bone morphogenetic protein 1 to its substrate chordin, and as a vascular tissue remodeler with pro-angiogenic properties. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2017].
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLFML3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OLFML3
Gene Accession No. B0BNI5, Q9NRN5
Gene Alias 2810002E22Rik; AI115209; AW550633; HNOEL-iso; hOLF44; mONT3; OLF44; olfactomedin like 3; olfactomedin-like 3; olfactomedin-like protein 3; OLFML3; ONT3; PSEC0035; PSEC0173; PSEC0244; UNQ663/PRO1294
Gene Symbols OLFML3
Host Species Rabbit
Immunogen Recombinant Human Olfactomedin-like protein 3 protein (201-406AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310743, 56944
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.