Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Olfr640 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524135
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524135 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524135 Supplier Invitrogen™ Supplier No. PA524135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Olfr640 Polyclonal Antibody for Western Blot

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olfr640
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR51I1
Gene Accession No. Q9H343
Gene Alias GA_x6K02T2PBJ9-6756759-6755815; HOR5'Beta11; MOR13-4; Odorant receptor HOR5'beta11; olfactory receptor; olfactory receptor 51I1; olfactory receptor 640; olfactory receptor family 51 subfamily I member 1; olfactory receptor MOR13-4; olfactory receptor OR11-39; olfactory receptor, family 51, subfamily I, member 1; Olfr640; OR51I1
Gene Symbols Olfr640, OR51I1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 284-313 amino acids from the C-terminal region of human OR51I1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 258819, 390063
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.