Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLIG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569395
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569395 Supplier Invitrogen™ Supplier No. PA569395
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

OLIG1 (oligodendrocyte transcription factor 1) promotes formation and maturation of oligodendrocytes - especially in the brain. It cooperates with OLIG2 to establish the pMN domain of the embryonic neural tube.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLIG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OLIG1
Gene Accession No. Q9WUQ3
Gene Alias AI836478; AW494459; basic domain, helix-loop-helix protein, class B, 6; BHLHB6; bHLHb7; bHLHe20; bHLHe21; class B basic helix-loop-helix protein 6; Class E basic helix-loop-helix protein 21; LOW QUALITY PROTEIN: oligodendrocyte transcription factor 1; Olg1; Olg-1; Olg-1 bHLH protein; OLIG1; olig2; olig3; Oligo1; Oligo2 antibody; oligodendrocyte lineage transcription factor 1; oligodendrocyte transcription factor 1; Oligodendrocyte-specific bHLH transcription factor 1
Gene Symbols OLIG1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human OLIG1 (aa 96-145).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60394
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.