Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olig1 Rabbit anti-Human, Mouse, Rat, Clone: 7E4B2, Novus Biologicals™
SDP

Catalog No. p-7246864 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165749 20 μg
NB165748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165749 Supplier Novus Biologicals Supplier No. NBP31532620UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Olig1 Monoclonal antibody specifically detects Olig1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Olig1
Applications Western Blot
Classification Monoclonal
Clone 7E4B2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias bHLHb6, BHLHE21, Class B basic helix-loop-helix protein 6, Class E basic helix-loop-helix protein 21, helix-loop-helix protein, class B, 6, oligo1, oligodendrocyte lineage transcription factor 1, oligodendrocyte transcription factor 1, oligodendrocyte-specific bHLH transcription factor 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 172-271 of human Olig1 (Q8TAK6).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 116448
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.