Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLIG1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8548R

Description
OLIG1 (oligodendrocyte transcription factor 1) promotes formation and maturation of oligodendrocytes - especially in the brain. It cooperates with OLIG2 to establish the pMN domain of the embryonic neural tube.Specifications
OLIG1 | |
Polyclonal | |
Unconjugated | |
OLIG1 | |
AI836478; AW494459; basic domain, helix-loop-helix protein, class B, 6; BHLHB6; bHLHb7; bHLHe20; BHLHE21; BOS_531; Class B basic helix-loop-helix protein 6; Class E basic helix-loop-helix protein 21; LOC100339501; LOW QUALITY PROTEIN: oligodendrocyte transcription factor 1; Olg1; Olg-1; Olg-1 bHLH protein; Olig1; olig2; olig3; Oligo1; Oligo2 antibody; oligodendrocyte lineage transcription factor 1; Oligodendrocyte transcription factor 1; oligodendrocyte transcription factor 1 (predicted)-like; Oligodendrocyte-specific bHLH transcription factor 1 | |
Rabbit | |
Protein A | |
RUO | |
116448 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8TAK6 | |
OLIG1 | |
KLH conjugated synthetic peptide derived from human OLIG1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction