Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Olig2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$192.47 - $467.51
Specifications
| Antigen | Olig2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400 - 1:800 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227301SS
![]() |
Novus Biologicals
NBP227301SS |
0.025 mg |
Each for $192.47
|
|
|||||
NBP227301
![]() |
Novus Biologicals
NBP227301 |
0.1 mg |
Each for $467.51
|
|
|||||
Description
Olig2 Polyclonal specifically detects Olig2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Olig2 | |
| Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400 - 1:800 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Q13516 | |
| 10215 | |
| A portion of amino acids 70-130 of human OLIG2 was used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuronal Stem Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| PBS with 0.05% Sodium Azide | |
| BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17helix-loop-helix protein, class B, 1 | |
| OLIG2 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for the antibody is 94% homologous in mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title