Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Olig2 Rabbit anti-Human, Mouse, Rat, Biotin, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc OLIG2BIOTIN

Description
Olig2 Polyclonal antibody specifically detects Olig2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Olig2 | |
Polyclonal | |
Biotin | |
OLIG2 | |
AI604895; basic domain, helix-loop-helix protein, class B, 1; bHLH transcription factor; BHLHB1; bHLHe19; Class B basic helix-loop-helix protein 1; class E basic helix-loop-helix protein 19; human protein kinase C-binding protein RACK17; Olg-2; OLIG2; olig-2; Oligo2; Oligo2 antibody; oligodendrocyte lineage transcription factor 2; oligodendrocyte transcription factor 2; oligodendrocyte-specific bHLH transcription factor 2; PRKCBP2; Protein kinase C-binding protein 2; Protein kinase C-binding protein RACK17; RACK17; RK17; wu:fc64g09 | |
Synthetic peptide taken within amino acid region 1-50 on human Olig2 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q13516, Q9EQW6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10215, 304103, 50913 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction