Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Olig3 Antibody (SR1487), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322426100UL
Description
Olig3 Monoclonal antibody specifically detects Olig3 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Olig3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
SR1487 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Bhlhb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3 | |
A synthesized peptide derived from human Olig3 (Uniprot #: Q7RTU3) | |
100 μg | |
Neuroscience | |
167826 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction