Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLIG3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501979976
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9976 Supplier Bioss Supplier No. BS20543R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OLIG3 may determine the distinct specification program of class A neurons in the dorsal part of the spinal cord and suppress specification of class B neurons.

Specifications

Antigen OLIG3
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 15mM sodium azide; pH
Gene OLIG3
Gene Accession No. Q7RTU3
Gene Alias basic helix-loop-helix domain-containing class B protein 7; Bhlhb7; BHLHE20; bHLHe20oligo3; Class B basic helix-loop-helix protein 7; Class E basic helix-loop-helix protein 20; LOC100349761; Olig3; Oligo3; oligodendrocyte transcription factor 2; Oligodendrocyte transcription factor 3; Oligodendrocyte-specific bHLH transcription factor 3
Gene Symbols OLIG3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OLIG3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167826
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.