Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OLIG3 Recombinant Rabbit Monoclonal Antibody (9C1N3)
GREENER_CHOICE

Catalog No. PIMA542609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542609 Supplier Invitrogen™ Supplier No. MA542609
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: U-251MG. The target is usually found in the following locations: Nucleus. Immunogen sequence: HAANSVHPVH PILGGALSSG NASSPLSAAS LPAIGTIRPP HSLLKAPSTP PALQLGSGFQ HWAGLPCPCT ICQMPPPPHL SALSTANMAR LSAESKDLLK.

Oligodendrocyte transcription factor 3 (OLIG3) is a gene located on chromosome 6q23.2, encoding a basic helix-loop-helix (bHLH) transcription factor that is involved in the regulation of neural development. OLIG3 is a member of the OLIG family, which plays a significant role in the differentiation and maturation of oligodendrocytes, the myelinating cells of the central nervous system. This transcription factor is crucial for the specification and development of certain neuronal subtypes, contributing to the formation and maintenance of the central nervous system's architecture. OLIG3's activity influences gene expression related to neural proliferation, survival, and differentiation. Although less studied than OLIG1 and OLIG2, OLIG3's expression patterns suggest its involvement in neural patterning and potential roles in neuronal disorders. Research continues to explore the mechanisms through which OLIG3 affects neural development and its implications in neuropathologies and regenerative medicine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OLIG3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9C1N3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OLIG3
Gene Accession No. Q7RTU3
Gene Alias basic helix-loop-helix domain-containing class B protein 7; Bhlhb7; bHLHe20; bHLHe20oligo3; Class B basic helix-loop-helix protein 7; class E basic helix-loop-helix protein 20; OLIG3; Oligo3; oligodendrocyte transcription factor 2; oligodendrocyte transcription factor 2; oligodendrocyte transcription factor 3; oligodendrocyte transcription factor 3; Oligodendrocyte-specific bHLH transcription factor 3
Gene Symbols OLIG3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 173-272 of human Olig3 (Q7RTU3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167826
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.