Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ OLIGO™ R3 Reversed Phase Columns

Catalog No. 1133426
Change view
Click to view available options
Material:
PEEK
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-334-26 Stainless Steel
11-332-26 PEEK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-334-26 Supplier Thermo Scientific™ Supplier No. 1133426
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.

Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.

Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:

  • High performance chromatography resin for analytical to process scale separations
  • High resolution preparative purification at high flow rates
  • Chemical stability—allows aggressive cleaning and sanitization
  • Poly-divinylbenzene particles—yield a robust, easily packable matrix
  • Combines low back pressure, high pH stability, and high capacity

Specifications

Column Format Prepacked Column
Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Max. Pressure 2400 psi (170 bar)
Particle Size 30 μm
Pore Size 350 Å
Quantity 1 Column
Stationary Phase Phenyl
Product Line POROS
Shipping Condition Room Temperature
Volume (Metric) 1.7 mL
Column Type Reversed Phase
Phase Reversed Phase
Material Stainless Steel
Technique Reversed Phase HPLC
Product Type Analytical Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.